C12H316CED05 Chemcia Scientific, LLC

Indazole-1,6-dicarboxylic acid 1-tert-butyl ester 6-methyl ester

  • MDL number MFCD170160~5C1
  • Cas Number : 1126424-50-7
  • Molecular Weight : 276.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=4181

Safety Information

msds http://www.chemcia.com/product_fl02.asp?id=4181
Availability details for
Quantity
  • Availability
  • Add to Cart