C12H316CED12 Chemcia Scientific, LLC

5-Chloro-[1,2,4]triazolo[1,5-a]pyrazin-2-ylamine

  • MDL number MFCD27634960
  • Cas Number : 1454653-85-0
  • Molecular Weight : 169.57413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4286
Availability details for
Quantity
  • Availability
  • Add to Cart