C12H316CED14 Chemcia Scientific, LLC

6-Fluoro-3-nitro-imidazo[1,2-a]pyridine

  • MDL number MFCD20483121
  • Cas Number : 1235993-30-2
  • Molecular Weight : 181.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-3-nitroimidazo[1,2-a]pyridine
Inchi Code 1S/C7H4FN3O2/c8-5-1-2-6-9-3-7(11(12)13)10(6)4-5/h1-4H
Inchi Key WTEGDZZHPUPJCU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4288

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4288
Availability details for
Quantity
  • Availability
  • Add to Cart