C12H316CED28 Chemcia Scientific, LLC

5-Bromo-4-chloro-2-fluoro-benzaldehyde

  • MDL number MFCD20486523
  • Cas Number : 1781052-25-2
  • Molecular Weight : 237.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-chloro-2-fluorobenzaldehyde
Inchi Code 1S/C7H3BrClFO/c8-5-1-4(3-11)7(10)2-6(5)9/h1-3H
Inchi Key XXIYYWQCGYDEFE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4319

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4319
Availability details for
Quantity
  • Availability
  • Add to Cart