C12H316CED39 Chemcia Scientific, LLC

1-(2,2-Dimethoxy-ethyl)-2-nitro-benzene

  • MDL number MFCD16890134
  • Cas Number : 79844-33-0
  • Molecular Weight : 211.21941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4440

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4440
Availability details for
Quantity
  • Availability
  • Add to Cart