C12H316CED58 Chemcia Scientific, LLC

2-Phenyl-2,5,6,7-tetrahydro-pyrazolo[4,3-b]pyridine-4-carboxylic acid tert-butyl ester

  • MDL number MFCD28348665
  • Cas Number : 1951439-79-4
  • Molecular Weight : 299.37582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4433

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4433
Availability details for
Quantity
  • Availability
  • Add to Cart