C12H316CED5C Chemcia Scientific, LLC

5-Bromo-4-chloro-2-fluoro-N-methoxy-N-methyl-benzamide

  • MDL number MFCD28348667
  • Cas Number : 1704367-16-7
  • Molecular Weight : 296.52501

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4438

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4438
Availability details for
Quantity
  • Availability
  • Add to Cart