C12H316CED5F Chemcia Scientific, LLC

(5-Bromo-3-nitro-pyridin-2-yl)-hydrazine

  • MDL number MFCD09801053
  • Cas Number : 15862-42-7
  • Molecular Weight : 233.0252

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Reddish Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4443

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4443
Availability details for
Quantity
  • Availability
  • Add to Cart