C12H316CED61 Chemcia Scientific, LLC

7-Bromo-1H-pyrido[2,3-b][1,4]thiazin-2-one

  • MDL number MFCD02930636
  • Cas Number : 438622-20-9
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-1H-pyrido[2,3-b][1,4]thiazin-2(3H)-one
Inchi Code 1S/C7H5BrN2OS/c8-4-1-5-7(9-2-4)12-3-6(11)10-5/h1-2H,3H2,(H,10,11)
Inchi Key DCCDHDKETKOVBR-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4445

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4445
Availability details for
Quantity
  • Availability
  • Add to Cart