C12H316CED62 Chemcia Scientific, LLC

7-Bromo-2,3-dihydro-1H-pyrido[2,3-b][1,4]thiazine

  • MDL number MFCD28348668
  • Cas Number : 1593552-66-9
  • Molecular Weight : 231.11524

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4447

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4447
Availability details for
Quantity
  • Availability
  • Add to Cart