C12H316CED65 Chemcia Scientific, LLC

3-Oxo-2,6,7,8-tetrahydro-3H-pyrido[3,2-c]pyridazine-5-carboxylic acid benzyl ester

  • MDL number MFCD28348669
  • Cas Number : 1936609-12-9
  • Molecular Weight : 285.3051

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4450

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4450
Availability details for
Quantity
  • Availability
  • Add to Cart