C12H316CED68 Chemcia Scientific, LLC

2-Dimethylaminomethylene-3-oxo-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD28348672
  • Cas Number : 1951451-51-6
  • Molecular Weight : 288.3494

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4452

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4452
Availability details for
Quantity
  • Availability
  • Add to Cart