C12H316CED90 Chemcia Scientific, LLC

1,5,6,7-Tetrahydro-pyrazolo[4,3-b]pyridine-4-carboxylic acid benzyl ester

  • MDL number MFCD28348688
  • Cas Number : 1935393-12-6
  • Molecular Weight : 257.29455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4592

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4592
Availability details for
Quantity
  • Availability
  • Add to Cart