C12H316CED91 Chemcia Scientific, LLC

2-Chloro-5-methoxy-4-nitro-benzoic acid

  • MDL number MFCD26395066
  • Cas Number : 101581-13-9
  • Molecular Weight : 231.59372

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4593

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4593
Availability details for
Quantity
  • Availability
  • Add to Cart