C12H316CED95 Chemcia Scientific, LLC

(3-Bromo-5-methyl-pyridin-2-yl)-methyl-amine

  • MDL number MFCD21911667
  • Cas Number : 872492-59-6
  • Molecular Weight : 201.06718

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4596

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4596
Availability details for
Quantity
  • Availability
  • Add to Cart