C12H316CED99 Chemcia Scientific, LLC

5-Bromo-2-chloro-6-methyl-pyridine-3-carbaldehyde

  • MDL number MFCD28348691
  • Cas Number : 1935331-10-4
  • Molecular Weight : 234.481

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4610

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4610
Availability details for
Quantity
  • Availability
  • Add to Cart