C12H316CED9A Chemcia Scientific, LLC

2,8-Dichloro-[1,2,4]triazolo[1,5-a]pyrazine

  • MDL number MFCD28348692
  • Cas Number : 1936295-68-9
  • Molecular Weight : 189.00449

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4585

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4585
Availability details for
Quantity
  • Availability
  • Add to Cart