C12H316CED9B Chemcia Scientific, LLC

6-Methyl-3-nitro-imidazo[1,2-a]pyridine

  • MDL number MFCD06496196
  • Cas Number : 67625-28-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-3-nitroimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7N3O2/c1-6-2-3-7-9-4-8(11(12)13)10(7)5-6/h2-5H,1H3
Inchi Key CUUHOYDMDHFMIY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4586

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4586
Availability details for
Quantity
  • Availability
  • Add to Cart