C12H316CED9C Chemcia Scientific, LLC

6-Methoxy-imidazo[1,2-b]pyridazine

  • MDL number MFCD12913581
  • Cas Number : 17240-33-4
  • Molecular Weight : 149.15334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4600

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4600
Availability details for
Quantity
  • Availability
  • Add to Cart