C12H316CED9E Chemcia Scientific, LLC

4-Bromo-2-chloro-5-nitro-benzoic acid methyl ester

  • MDL number MFCD12032200
  • Cas Number : 1157672-86-0
  • Molecular Weight : 294.49035

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4602

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4602
Availability details for
Quantity
  • Availability
  • Add to Cart