C12H316CEDA4 Chemcia Scientific, LLC

6-Chloro-2H-indazol-3-ylamine

  • MDL number MFCD20265440
  • Cas Number : 41449-95-0
  • Molecular Weight : 167.59897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4607

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4607
Availability details for
Quantity
  • Availability
  • Add to Cart