C12H316CEDA7 Chemcia Scientific, LLC

1-Chloro-4-methoxy-2-methyl-5-nitro-benzene

  • MDL number MFCD01861454
  • Cas Number : 101080-03-9
  • Molecular Weight : 201.61086

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4612

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4612
Availability details for
Quantity
  • Availability
  • Add to Cart