C12H316CEDA8 Chemcia Scientific, LLC

(5-Bromo-2-chloro-6-methyl-pyridin-3-yl)-methanol

  • MDL number MFCD28348697
  • Cas Number : 1936227-62-1
  • Molecular Weight : 236.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-bromo-2-chloro-6-methylpyridin-3-yl)methanol
Inchi Code 1S/C7H7BrClNO/c1-4-6(8)2-5(3-11)7(9)10-4/h2,11H,3H2,1H3
Inchi Key HFYQEIXPNYSTRL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4609

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4609
Availability details for
Quantity
  • Availability
  • Add to Cart