C12H316CEDAA Chemcia Scientific, LLC

2-Fluoro-4-methyl-5-nitro-benzoic acid

  • MDL number MFCD18426005
  • Cas Number : 753924-40-2
  • Molecular Weight : 199.13972

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4614

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4614
Availability details for
Quantity
  • Availability
  • Add to Cart