C12H316CEDAB Chemcia Scientific, LLC

2-Fluoro-4-methyl-5-nitro-benzoic acid methyl ester

  • MDL number MFCD23863743
  • Cas Number : 753924-48-0
  • Molecular Weight : 213.16681

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4615

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4615
Availability details for
Quantity
  • Availability
  • Add to Cart