C12H316CEDAD Chemcia Scientific, LLC

5-Chloro-2-fluoro-4-methyl-phenylamine

  • MDL number MFCD18426015
  • Cas Number : 1263275-21-3
  • Molecular Weight : 159.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-chloro-2-fluoro-4-methylaniline
Inchi Code 1S/C7H7ClFN/c1-4-2-6(9)7(10)3-5(4)8/h2-3H,10H2,1H3
Inchi Key QLSFSYFWNWRGLG-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4617

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4617
Availability details for
Quantity
  • Availability
  • Add to Cart