C12H316CEDAE Chemcia Scientific, LLC

2,4,5,7-Tetrahydro-pyrazolo[3,4-c]pyridine-6-carboxylic acid benzyl ester

  • MDL number MFCD28348698
  • Cas Number : 1934668-58-2
  • Molecular Weight : 257.29455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4588

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4588
Availability details for
Quantity
  • Availability
  • Add to Cart