C12H316CEDAF Chemcia Scientific, LLC

1,4,5,7-Tetrahydro-pyrazolo[3,4-c]pyridine-6-carboxylic acid benzyl ester

  • MDL number MFCD28348699
  • Cas Number : 1934668-58-2
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 1,4,5,7-tetrahydro-6H-pyrazolo[3,4-c]pyridine-6-carboxylate
Inchi Code 1S/C14H15N3O2/c18-14(19-10-11-4-2-1-3-5-11)17-7-6-12-8-15-16-13(12)9-17/h1-5,8H,6-7,9-10H2,(H,15,16)
Inchi Key NQEGFSHMUOFDQX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4589

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4589
Availability details for
Quantity
  • Availability
  • Add to Cart