C12H316CEDD6 Chemcia Scientific, LLC

7-Methyl-3-nitro-imidazo[1,2-a]pyridine

  • MDL number MFCD00179132
  • Cas Number : 34165-07-6
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-methyl-3-nitroimidazo[1,2-a]pyridine
Inchi Code 1S/C8H7N3O2/c1-6-2-3-10-7(4-6)9-5-8(10)11(12)13/h2-5H,1H3
Inchi Key ZOAYHVZFPBPFJI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4814

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4814
Availability details for
Quantity
  • Availability
  • Add to Cart