C12H316CEDD9 Chemcia Scientific, LLC

6-Methyl-benzo[d]isoxazol-3-ylamine

  • MDL number MFCD18803474
  • Cas Number : 1378699-89-8
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methyl-1,2-benzisoxazol-3-amine
Inchi Code 1S/C8H8N2O/c1-5-2-3-6-7(4-5)11-10-8(6)9/h2-4H,1H3,(H2,9,10)
Inchi Key AXUHXLAVQYXDMT-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4817

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4817
Availability details for
Quantity
  • Availability
  • Add to Cart