C12H316CEDDB Chemcia Scientific, LLC

1-Bromo-4-tert-butyl-2-nitro-benzene

  • MDL number MFCD11845934
  • Cas Number : 70729-05-4
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-4-tert-butyl-2-nitrobenzene
Inchi Code 1S/C10H12BrNO2/c1-10(2,3)7-4-5-8(11)9(6-7)12(13)14/h4-6H,1-3H3
Inchi Key XACFLGSMDUITKB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4819

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4819
Availability details for
Quantity
  • Availability
  • Add to Cart