C12H316CEDDC Chemcia Scientific, LLC

(4-Chloro-2-fluoro-5-iodo-phenyl)-methanol

  • MDL number MFCD28400708
  • Cas Number : 1935084-33-5
  • Molecular Weight : 286.4731

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4820

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4820
Availability details for
Quantity
  • Availability
  • Add to Cart