C12H316CEDE0 Chemcia Scientific, LLC

5-Iodo-6-methoxy-3-nitro-pyridin-2-ylamine

  • MDL number MFCD17014969
  • Cas Number : 868539-54-2
  • Molecular Weight : 295.03742

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4825

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4825
Availability details for
Quantity
  • Availability
  • Add to Cart