C12H316CEDE3 Chemcia Scientific, LLC

(4,5-Difluoro-2-nitro-phenyl)-methanol

  • MDL number MFCD20485905
  • Cas Number : 914095-13-9
  • Molecular Weight : 189.1196

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4828

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4828
Availability details for
Quantity
  • Availability
  • Add to Cart