C12H316CEDF6 Chemcia Scientific, LLC

2-Bromo-4-thiocyanato-phenylamine

  • MDL number MFCD04618254
  • Cas Number : 7493-98-3
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-amino-3-bromophenyl thiocyanate
Inchi Code 1S/C7H5BrN2S/c8-6-3-5(11-4-9)1-2-7(6)10/h1-3H,10H2
Inchi Key UUQUVDNHTRZTCV-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4785

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4785
Availability details for
Quantity
  • Availability
  • Add to Cart