C12H316CEDF7 Chemcia Scientific, LLC

2-Bromo-1-chloro-4-thiocyanato-benzene

  • Molecular Weight : 248.53

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-chloro-4-thiocyanatobenzene
Inchi Code 1S/C7H3BrClNS/c8-6-3-5(11-4-10)1-2-7(6)9/h1-3H
Inchi Key ZJZCCDZDGHDGJA-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Brownish Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4786

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4786
Availability details for
Quantity
  • Availability
  • Add to Cart