C12H316CEDFC Chemcia Scientific, LLC

4-Fluoro-5-hydroxy-2-nitro-benzoic acid

  • MDL number MFCD16159128
  • Cas Number : 38569-85-6
  • Molecular Weight : 201.11203

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4833

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4833
Availability details for
Quantity
  • Availability
  • Add to Cart