C12H316CEDFF Chemcia Scientific, LLC

4-Fluoro-5-hydroxy-2-nitro-benzoic acid methyl ester

  • MDL number MFCD23699495
  • Cas Number : 1426958-40-8
  • Molecular Weight : 215.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-fluoro-5-hydroxy-2-nitrobenzoate
Inchi Code 1S/C8H6FNO5/c1-15-8(12)4-2-7(11)5(9)3-6(4)10(13)14/h2-3,11H,1H3
Inchi Key IWYPVZJXNKHKRF-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4837

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4837
Availability details for
Quantity
  • Availability
  • Add to Cart