C12H316CEE00 Chemcia Scientific, LLC

2-Amino-4,5-difluoro-benzonitrile

  • MDL number MFCD09835282
  • Cas Number : 219823-49-1
  • Molecular Weight : 154.12013

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Tan Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4838

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4838
Availability details for
Quantity
  • Availability
  • Add to Cart