C12H316CEE03 Chemcia Scientific, LLC

2-Amino-4-fluoro-5-hydroxy-benzoic acid methyl ester

  • MDL number MFCD16159183
  • Cas Number : 1113049-61-8
  • Molecular Weight : 185.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-4-fluoro-5-hydroxybenzoate
Inchi Code 1S/C8H8FNO3/c1-13-8(12)4-2-7(11)5(9)3-6(4)10/h2-3,11H,10H2,1H3
Inchi Key FSUONMUAAVMIJC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4841

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4841
Availability details for
Quantity
  • Availability
  • Add to Cart