C12H316CEE07 Chemcia Scientific, LLC

6-Iodo-benzo[d]isoxazol-3-ylamine

  • Cas Number : 1260902-18-8
  • Molecular Weight : 260.0351

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4794

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4794
Availability details for
Quantity
  • Availability
  • Add to Cart