C12H316CEE09 Chemcia Scientific, LLC

6-Iodo-2H-indazol-3-ylamine

  • Molecular Weight : 259.05037

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4796

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4796
Availability details for
Quantity
  • Availability
  • Add to Cart