C12H316CEE0C Chemcia Scientific, LLC

6-Bromo-3-iodo-imidazo[1,2-b]pyridazine

  • Cas Number : 1609565-17-4
  • Molecular Weight : 323.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-3-iodoimidazo[1,2-b]pyridazine
Inchi Code 1S/C6H3BrIN3/c7-4-1-2-6-9-3-5(8)11(6)10-4/h1-3H
Inchi Key XDMYPVQOFKSMRJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4811

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4811
Availability details for
Quantity
  • Availability
  • Add to Cart