C12H316CEE0E Chemcia Scientific, LLC

6-Iodo-3-nitro-pyridin-2-ylamine

  • MDL number MFCD27975897
  • Cas Number : 351447-13-7
  • Molecular Weight : 265.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodo-3-nitropyridin-2-amine
Inchi Code 1S/C5H4IN3O2/c6-4-2-1-3(9(10)11)5(7)8-4/h1-2H,(H2,7,8)
Inchi Key OFNGNLQJHMFYLB-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4836

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4836
Availability details for
Quantity
  • Availability
  • Add to Cart