C12H316CEE0F Chemcia Scientific, LLC

3,5-Diiodo-2-methoxy-benzonitrile

  • MDL number MFCD28400722
  • Cas Number : 898731-72-1
  • Molecular Weight : 384.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-diiodo-2-methoxybenzonitrile
Inchi Code 1S/C8H5I2NO/c1-12-8-5(4-11)2-6(9)3-7(8)10/h2-3H,1H3
Inchi Key CIUAETIYCHXSBK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4843

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4843
Availability details for
Quantity
  • Availability
  • Add to Cart