C12H316CEE10 Chemcia Scientific, LLC

8-Fluoro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD13195311
  • Cas Number : 1260843-88-6
  • Molecular Weight : 208.19383

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4844

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4844
Availability details for
Quantity
  • Availability
  • Add to Cart