C12H316CEE11 Chemcia Scientific, LLC

2-Hydroxy-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD17170446
  • Cas Number : 69622-67-9
  • Molecular Weight : 235.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name benzyl 2-hydroxy-1-piperidinecarboxylate
Inchi Code 1S/C13H17NO3/c15-12-8-4-5-9-14(12)13(16)17-10-11-6-2-1-3-7-11/h1-3,6-7,12,15H,4-5,8-10H2
Inchi Key DJHBQZYPXJJJMZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4845

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4845
Availability details for
Quantity
  • Availability
  • Add to Cart