C12H316CEE17 Chemcia Scientific, LLC

8-Fluoro-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09994934
  • Cas Number : 1020034-56-3
  • Molecular Weight : 180.13965

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4854

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4854
Availability details for
Quantity
  • Availability
  • Add to Cart