C12H316CEE19 Chemcia Scientific, LLC

3,5-Dibromo-1H-indazole

  • MDL number MFCD00159919
  • Cas Number : 40598-76-3
  • Molecular Weight : 275.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,5-dibromo-1H-indazole
Inchi Code 1S/C7H4Br2N2/c8-4-1-2-6-5(3-4)7(9)11-10-6/h1-3H,(H,10,11)
Inchi Key YPNFVZZHYXRCKH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4856

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4856
Availability details for
Quantity
  • Availability
  • Add to Cart