C12H316CEE1D Chemcia Scientific, LLC

8-Fluoro-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD18072717
  • Cas Number : 1245644-74-9
  • Molecular Weight : 152.13195

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4861

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4861
Availability details for
Quantity
  • Availability
  • Add to Cart